Merck
  • Notch signaling plays a critical role in motility and differentiation of human first-trimester cytotrophoblasts.
Notch signaling plays a critical role in motility and differentiation of human first-trimester cytotrophoblasts.
Endocrinology (2013-11-06)
Sandra Haider, Gudrun Meinhardt, Philipp Velicky, Gerlinde R Otti, Guy Whitley, Christian Fiala, Jürgen Pollheimer, Martin Knöfler
ABSTRACT
Failures in human extravillous trophoblast (EVT) development could be involved in the pathogenesis of pregnancy diseases. However, the underlying mechanisms have been poorly characterized. Here, we provide evidence that Notch signaling could represent a key regulatory pathway controlling trophoblast proliferation, motility, and differentiation. Immunofluorescence of first-trimester placental tissues revealed expression of Notch receptors (Notch2 and Notch3) and membrane-anchored ligands (delta-like ligand [DLL] 1 and -4 and Jagged [JAG] 1 and -2) in villous cytotrophoblasts (vCTBs), cell column trophoblasts (CCTs), and EVTs. Notch4 and Notch1 were exclusively expressed in vCTBs and in CCTs, respectively. Both proteins decreased in Western blot analyses of first-trimester, primary cytotrophoblasts (CTBs) differentiating on fibronectin. Luciferase reporter analyses suggested basal, canonical Notch activity in SGHPL-5 cells and primary cells that was increased upon seeding on DLL4-coated dishes and diminished in the presence of the Notch/γ-secretase inhibitors N-[N-(3,5-difluorophenacetyl-l-alanyl)]-S-phenylglycine t-butyl ester (DAPT) or L-685,458. Bromodeoxyuridine labeling, cyclin D1 mRNA expression, and cell counting indicated that chemical inhibition of Notch signaling elevated proliferation in the different primary trophoblast model systems. Notch inhibition also increased motility of SGHPL-5 cells through uncoated and fibronectin-coated Transwells, motility of primary CTBs, as well as migration in villous explant cultures on collagen I. Accordingly, small interfering RNA-mediated gene silencing of Notch1 also elevated SGHPL-5 cell migration. In contrast, motility of primary cultures and SGHPL-5 cells was diminished in the presence of DLL4. Moreover, DAPT increased markers of differentiated EVT, ie, human leukocyte antigen G1, integrin α5, and T-cell factor 4, whereas DLL4 provoked the opposite. In summary, the data suggest that canonical Notch signaling impairs motility and differentiation of first-trimester CTBs.
Sigma-Aldrich
Anti-Ki-67 Antibody, clone Ki-S5, clone Ki-S5, Chemicon®, from mouse
Sigma-Aldrich
γ-Secretase Inhibitor IX, Gamma-Secretase Inhibitor IX - CAS 208255-80-5, is a cell-permeable inhibitor of γ-secretase (Aβtotal IC₅₀ = 115 nM, Aβ42 IC₅₀ = 200 nM).