Merck
  • Mitochondrial GTP Links Nutrient Sensing to β Cell Health, Mitochondrial Morphology, and Insulin Secretion Independent of OxPhos.
Mitochondrial GTP Links Nutrient Sensing to β Cell Health, Mitochondrial Morphology, and Insulin Secretion Independent of OxPhos.
Cell reports (2019-07-18)
Sean R Jesinkey, Anila K Madiraju, Tiago C Alves, OrLando H Yarborough, Rebecca L Cardone, Xiaojian Zhao, Yassmin Parsaei, Ali R Nasiri, Gina Butrico, Xinran Liu, Anthony J Molina, Austin M Rountree, Adam S Neal, Dane M Wolf, John Sterpka, William M Philbrick, Ian R Sweet, Orian H Shirihai, Richard G Kibbey
ABSTRACT
Mechanisms coordinating pancreatic β cell metabolism with insulin secretion are essential for glucose homeostasis. One key mechanism of β cell nutrient sensing uses the mitochondrial GTP (mtGTP) cycle. In this cycle, mtGTP synthesized by succinyl-CoA synthetase (SCS) is hydrolyzed via mitochondrial PEPCK (PEPCK-M) to make phosphoenolpyruvate, a high-energy metabolite that integrates TCA cycling and anaplerosis with glucose-stimulated insulin secretion (GSIS). Several strategies, including xenotopic overexpression of yeast mitochondrial GTP/GDP exchanger (GGC1) and human ATP and GTP-specific SCS isoforms, demonstrated the importance of the mtGTP cycle. These studies confirmed that mtGTP triggers and amplifies normal GSIS and rescues defects in GSIS both in vitro and in vivo. Increased mtGTP synthesis enhanced calcium oscillations during GSIS. mtGTP also augmented mitochondrial mass, increased insulin granule number, and membrane proximity without triggering de-differentiation or metabolic fragility. These data highlight the importance of the mtGTP signal in nutrient sensing, insulin secretion, mitochondrial maintenance, and β cell health.
Sigma-Aldrich
Glucokinase Activator, Cpd A, The Glucokinase Activator, Cpd A, also referenced under CAS 603108-44-7, modulates Glucokinase. This small molecule/inhibitor is primarily used for Activators/Inducers applications.
Sigma-Aldrich
Histopaque®-1119, sterile-filtered, density: 1.119
Sigma-Aldrich
Acridine, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-Guinea Pig IgG (H+L), highly cross-adsorbed, CF555 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution