Merck
  • Peroxisome-proliferator-activated receptors and the control of levels of prostaglandin-endoperoxide synthase 2 by arachidonic acid in the bovine uterus.
Peroxisome-proliferator-activated receptors and the control of levels of prostaglandin-endoperoxide synthase 2 by arachidonic acid in the bovine uterus.
The Biochemical journal (2007-05-23)
E Linda R Sheldrick, Kamila Derecka, Elaine Marshall, Evonne C Chin, Louise Hodges, D Claire Wathes, D Robert E Abayasekara, Anthony P F Flint
ABSTRACT
Arachidonic acid is a potential paracrine agent released by the uterine endometrial epithelium to induce PTGS2 [PG (prostaglandin)-endoperoxide synthase 2] in the stroma. In the present study, bovine endometrial stromal cells were used to determine whether PTGS2 is induced by arachidonic acid in stromal cells, and to investigate the potential role of PPARs (peroxisome-proliferator-activated receptors) in this effect. Arachidonic acid increased PTGS2 levels up to 7.5-fold within 6 h. The cells expressed PPARalpha and PPARdelta (also known as PPARbeta) (but not PPARgamma). PTGS2 protein level was increased by PPAR agonists, including polyunsaturated fatty acids, synthetic PPAR ligands, PGA1 and NSAIDs (non-steroidal anti-inflammatory drugs) with a time course resembling that of arachidonic acid. Use of agonists and antagonists indicated PPARalpha (but not PPARdelta or PPARgamma) was responsible for PTGS2 induction. PTGS2 induction by arachidonic acid did not require PG synthesis. PTGS2 levels were increased by the PKC (protein kinase C) activators 4beta-PMA and PGF(2alpha), and the effects of arachidonic acid, NSAIDs, synthetic PPAR ligands and 4beta-PMA were blocked by PKC inhibitors. This is consistent with PPAR phosphorylation by PKC. Induction of PTGS2 protein by 4beta-PMA in the absence of a PPAR ligand was decreased by the NF-kappaB (nuclear factor kappaB) inhibitors MG132 and parthenolide, suggesting that PKC acted through NF-kappaB in addition to PPAR phosphorylation. Use of NF-kappaB inhibitors allowed the action of arachidonic acid as a PPAR agonist to be dissociated from an effect through PKC. The results are consistent with the hypothesis that arachidonic acid acts via PPARalpha to increase PTGS2 levels in bovine endometrial stromal cells.
Sigma-Aldrich
Linoleic acid, conjugated