Merck
  • Specificity and mechanism of protein kinase C activation by sn-1,2-diacylglycerols.
Specificity and mechanism of protein kinase C activation by sn-1,2-diacylglycerols.
Proceedings of the National Academy of Sciences of the United States of America (1986-03-01)
B R Ganong, C R Loomis, Y A Hannun, R M Bell
ABSTRACT
The specificity of protein kinase C activation by sn-1,2-diacylglycerols and analogues was investigated by using a Triton X-100 mixed micellar assay [Hannun, Y. A., Loomis, C. R. & Bell, R. M. (1985) J. Biol. Chem. 260, 10039-10043]. Analogues containing acyl or alkyl chains eight carbons in length were synthesized because sn-1,2-dioctanoylglycerol is an effective cell-permeant activator of protein kinase C. These analogues were tested as activators and antagonists of rat brain protein kinase C to determine the exact structural features important for activity. The analogues established that activation of protein kinase C by diacylglycerols is highly specific. Several analogues established that both carbonyl moieties of the oxygen esters are required for maximal activity and that the 3-hydroxyl moiety is also required. None of the analogues were antagonists. These data, combined with previous investigations, permitted formulation of a model of protein kinase C activation. A three-point attachment of sn-1,2-diacylglycerol to the surface-bound protein kinase C-phosphatidylserine-Ca2+ complex is envisioned to cause activation. Direct ligation of diacylglycerol to Ca2+ is proposed to be an essential step in the mechanism of activation of protein kinase C.
Product Description
Avanti
16:0-18:1 DG, 1-palmitoyl-2-oleoyl-sn-glycerol, neat oil
Avanti
16:0 DG, 1,2-dipalmitoyl-sn-glycerol, powder
Avanti
18:1 DG, Avanti Research - A Croda Brand
Avanti
16:0-18:1 DG, 1-palmitoyl-2-oleoyl-sn-glycerol, chloroform
Avanti
16:0 DG, 1,2-dipalmitoyl-sn-glycerol, chloroform
Avanti
08:0 DG, 1,2-dioctanoyl-sn-glycerol, neat oil
Avanti
10:0 DG, 1,2-didecanoyl-sn-glycerol, powder
Avanti
18:1 DG, 1-2-dioleoyl-sn-glycerol, chloroform
Avanti
08:0 DG, 1,2-dioctanoyl-sn-glycerol, chloroform solution
Avanti
12:0 DG, Avanti Research - A Croda Brand
Avanti
14:0 DG, 1,2-dimyristoyl-sn-glycerol, powder