Merck
Select acetophenones modulate flagellar motility in chlamydomonas.
Chemical biology & drug design (2010-07-28)
Shakila K Evans, Austin A Pearce, Prudence K Ibezim, Todd P Primm, Anne R Gaillard
ABSTRACT
Acetophenones were screened for activity against positive phototaxis of Chlamydomonas cells, a process that requires co-ordinated flagellar motility. The structure-activity relationships of a series of acetophenones are reported, including acetophenones that affect flagellar motility and cell viability. Notably, 4-methoxyacetophenone, 3,4-dimethoxyacetophenone, and 4-hydroxyacetophenone induced negative phototaxis in Chlamydomonas, suggesting interference with activity of flagellar proteins and control of flagellar dominance.
Sigma-Aldrich
3′,4′-Dimethylacetophenone, 98%
Sigma-Aldrich
Acetanisole, ≥98%, FCC, FG
Sigma-Aldrich
4′-Hydroxyacetophenone, 99%
Sigma-Aldrich
4′-Methoxyacetophenone, 99%