Merck
  • Efficient delivery of a Bcl-2-specific antisense oligodeoxyribonucleotide (G3139) via transferrin receptor-targeted liposomes.
Efficient delivery of a Bcl-2-specific antisense oligodeoxyribonucleotide (G3139) via transferrin receptor-targeted liposomes.
Journal of controlled release : official journal of the Controlled Release Society (2006-03-28)
Shih-Jiuan Chiu, Shujun Liu, Danilo Perrotti, Guido Marcucci, Robert J Lee
ABSTRACT
A novel transferrin receptor (TfR)-targeted liposomal formulation was synthesized and evaluated for the delivery of a phosphorothioate antisense oligodeoxyribonucleotide (ODN) (G3139, oblimerson sodium, or Genasense) to Bcl-2 in K562 leukemia cells. Liposomes composed of DC-Chol/egg PC/PEG-DSPE (25:73.5:1.5, mol/mol/mol) were loaded with G3139 with high efficiency (70-80%). To prepare targeted liposomes, transferrin was first coupled to PEG-DSPE and then incorporated into the bilayer by post-insertion. The liposomes had a mean diameter of 100 to 150 nm and exhibited colloidal stability for up to 8 weeks. Uptake of Tf-conjugated G3139-containing liposomes in TfR positive K562 cells was found to be more efficient than that of the non-targeted control formulation and could be blocked by excess free Tf. Treatment with Tf-conjugated liposomes resulted in Bcl-2 protein downregulation in K562 cells that was approximately 2-fold greater than with non-targeted liposomes (p<0.05) and 10-fold greater than with free G3139. Treatment with 2 microM G3139 in Tf-conjugated liposomes resulted in >80% reduction in Bcl-2 transcript. In addition, Tf-conjugated liposomal G3139-sensitized K562 cells to daunorubicin, lowering IC50 from 1.8 microM to 0.18 microM. In conclusion, Tf-conjugated liposomes are effective delivery vehicles for G3139 antisense oligos in TfR positive K562 cells and warrant further investigation as an in vivo oligo delivery vehicle.
Product Description
Avanti
DSPE-PEG(5000) Folate, Avanti Research - A Croda Brand 880123P, powder
Avanti
DSPE-PEG(2000) Folate, Avanti Research - A Croda Brand 880124X
Avanti
DSPE-PEG(5000) Folate, Avanti Research - A Croda Brand 880123X
Avanti
DSPE-PEG(2000) Folate, Avanti Research - A Croda Brand 880124P, powder