Merck
  • Effect of dietary docosahexaenoic acid on rhodopsin content and packing in photoreceptor cell membranes.
Effect of dietary docosahexaenoic acid on rhodopsin content and packing in photoreceptor cell membranes.
Biochimica et biophysica acta (2018-04-08)
Subhadip Senapati, Megan Gragg, Ivy S Samuels, Vipul M Parmar, Akiko Maeda, Paul S-H Park
ABSTRACT
Docosahexaenoic acid (DHA) is enriched in photoreceptor cell membranes. DHA deficiency impairs vision due to photoreceptor cell dysfunction, which is caused, at least in part, by reduced activity of rhodopsin, the light receptor that initiates phototransduction. It is unclear how the depletion of membrane DHA impacts the structural properties of rhodopsin and, in turn, its activity. Atomic force microscopy (AFM) was used to assess the impact of DHA deficiency on membrane structure and rhodopsin organization. AFM revealed that signaling impairment in photoreceptor cells is independent of the oligomeric status of rhodopsin and causes adaptations in photoreceptor cells where the content and density of rhodopsin in the membrane is increased. Functional and structural changes caused by DHA deficiency were reversible.
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid-21,21,22,22,22-d5, ≥98 atom % D, ≥98% (CP)
Sigma-Aldrich
Cyclopentolate