
Z-His-OH

Synonym(s):
Nα-Carbobenzyloxy-L-histidine, Nα-Z-L-Histidine
Empirical Formula (Hill Notation):
C14H15N3O4
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing