
Trospium impurity A

Synonym(s):
Benzilic acid, Diphenylglycolic acid
Linear Formula:
(C6H5)2C(OH)COOH
CAS Number:
Molecular Weight:
228.24
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing