
Triton™ X-100

Synonym(s):
t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether, Triton™ X-100
Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:
Products
Product Category
Brand
Biological Source
Feature
Physical Form
Product Line
Quality Segment
Application
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing