Skip to Content
Merck
Theophylline Related Compound B United States Pharmacopeia (USP) Reference Standard

Theophylline Related Compound B

Theophylline Related Compound B United States Pharmacopeia (USP) Reference Standard
Synonym(s):
3-Methylxanthine, 2,6-Dihydroxy-3-methylpurine
Empirical Formula (Hill Notation):
C6H6N4O2
CAS Number:
Molecular Weight:
166.14

Products

Brand
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade