
Tetrabutylammonium tetraphenylborate

Synonym(s):
Tetraphenylboron tetrabutylammonium
Linear Formula:
(CH3CH2CH2CH2)4N[B(C6H5)4]
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing