
Sophoricoside

Synonym(s):
5′,7′-Dihydroxy-4′-glucosyloxyisoflavone, Genistein-4′-O-glucoside
Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing