
Sodium ascorbate

Synonym(s):
(+)-Sodium L-ascorbate, L(+)-Ascorbic acid sodium salt, Vitamin C sodium salt
Empirical Formula (Hill Notation):
C6H7NaO6
CAS Number:
Molecular Weight:
198.11
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique