
Residual Solvent Class 1 - 1,1-Dichloroethene

Synonym(s):
1,1-Dichloroethene solution
Empirical Formula (Hill Notation):
C2H2Cl2
CAS Number:
Molecular Weight:
96.94
Products
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing