
Pyrifenox

Empirical Formula (Hill Notation):
C14H12Cl2N2O
CAS Number:
Molecular Weight:
295.16
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing