
Pymetrozin Metabolite CGA313124

Synonym(s):
6-Hydroxymethyl-4-(pyridin-3-ylmethylenamino)-4,5-dihydro-1,2,4-triazin-3(2H)one
Empirical Formula (Hill Notation):
C10H11N5O2
Molecular Weight:
233.23
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing