
Pramipexole for system suitability

Synonym(s):
Pramipexole dihydrochloride monohydrate
Empirical Formula (Hill Notation):
C10H17N3S · 2HCl · H2O
CAS Number:
Molecular Weight:
302.26
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing