
Potassium tert-pentoxide solution

Synonym(s):
Potassium (2-methyl-2-butoxide), Potassium tert-pentylate
Linear Formula:
CH3CH2C(CH3)2OK
CAS Number:
Molecular Weight:
126.24
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing