
PIPES dipotassium salt

Synonym(s):
1,4-Piperazinediethanesulfonic acid dipotassium salt, Piperazine-1,4-bis(2-ethanesulfonic acid) dipotassium salt
Empirical Formula (Hill Notation):
C8H16K2N2O6S2
CAS Number:
Molecular Weight:
378.55
Products
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing