
Pinacolyl methylphosphonate

Linear Formula:
CH3P(O)(OH)OCH(CH3)C(CH3)3
CAS Number:
Molecular Weight:
180.18
Products
Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing