![Phenytoin for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/351/938/a176722c-d180-4a73-b24e-298fad6def1d/640/a176722c-d180-4a73-b24e-298fad6def1d.png)
Phenytoin for system suitability
![Phenytoin for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/351/938/a176722c-d180-4a73-b24e-298fad6def1d/640/a176722c-d180-4a73-b24e-298fad6def1d.png)
Synonym(s):
5,5-Diphenylhydantoin, 5,5-Diphenyl-2,4-imidazolidinedione, Phenytoin
Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing