
Phenylalanine Ammonia-Lyase from Rhodotorula glutinis

Synonym(s):
L-Phenylalanine ammonia-lyase, Phenylalanine Deaminase from Rhodotorula glutinis
CAS Number:
Products
Brand
Biological Source
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing