
Orphenadrine Related Compound B

Synonym(s):
N-Ethyl-N,N-dimethyl [2-(2-methylbenzhydryloxy)ethyl]ammonium chloride
Empirical Formula (Hill Notation):
C20H28ClNO
CAS Number:
Molecular Weight:
333.90
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2