Skip to Content
Merck
Orphenadrine hydrochloride European Pharmacopoeia (EP) Reference Standard

Orphenadrine hydrochloride

Orphenadrine hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
β-Dimethylaminoethyl 2-methylbenzhydryl ether hydrochloride, N,N-Dimethyl-2-(2-methylbenzhydryloxy)ethylamine hydrochloride, o-Methyldiphenhydramine hydrochloride
Empirical Formula (Hill Notation):
C18H23NO · HCl
CAS Number:
Molecular Weight:
305.84

Products

Product Category
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2