Skip to Content
Merck
N-Methoxy-N-methyl-2,2,2-trifluoroacetamide 98%

N-Methoxy-N-methyl-2,2,2-trifluoroacetamide

N-Methoxy-N-methyl-2,2,2-trifluoroacetamide 98%
Synonym(s):
N-Methyl-N-methoxytrifluoroacetamide
Empirical Formula (Hill Notation):
C4H6F3NO2
CAS Number:
Molecular Weight:
157.09

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing