Skip to Content
Merck
Neocuproine hydrochloride hydrate BioReagent, crystalline

Neocuproine hydrochloride hydrate

Neocuproine hydrochloride hydrate BioReagent, crystalline
Synonym(s):
2,9-Dimethyl-1,10-phenanthroline hydrochloride hydrate
Empirical Formula (Hill Notation):
C14H12N2 · HCl · xH2O
CAS Number:
Molecular Weight:
244.72 (anhydrous basis)
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing