
Naltrexone

Synonym(s):
(5α)-17-(Cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxymorphinan-6-one, Naltrexone
Empirical Formula (Hill Notation):
C20H23NO4
CAS Number:
Molecular Weight:
341.40
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing