
Methylmalonyl coenzyme A tetralithium salt hydrate

Synonym(s):
α-Methylmalonyl coenzyme A tetralithium salt, MM-CoA
Empirical Formula (Hill Notation):
C25H36Li4N7O19P3S · xH2O
CAS Number:
Molecular Weight:
891.34 (anhydrous basis)
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing