
Mepyramine impurity A

Synonym(s):
2-(4-Methoxybenzylamino)pyridine
Empirical Formula (Hill Notation):
C13H14N2O
CAS Number:
Molecular Weight:
214.26
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing