Mebendazole-amine
Synonym(s):
2-Amino-5-benzoyl-1H-benzimidazole
Empirical Formula (Hill Notation):
C14H11N3O
CAS Number:
Molecular Weight:
237.26
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing