
Lisinopril Impurity A

Synonym(s):
DL-Homophenylalanine, (±)-2-Amino-4-phenylbutyric acid
Linear Formula:
C6H5(CH2CH2)CH(NH2)CO2H
CAS Number:
Molecular Weight:
179.22
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing