
Linezolid Related Compound B

Synonym(s):
N-[[(5S)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]ethanethioamide, (S)-N-{[3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl]methyl}thioacetamide
Empirical Formula (Hill Notation):
C16H20FN3O3S
CAS Number:
Molecular Weight:
353.41
Products
Brand
Formula Weight
Manufacturer
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2