
Kasugamycin hydrochloride from Streptomyces kasugaensis

Synonym(s):
3-O-[2-Amino-4-[(carboxyiminomethyl)amino]-2,3,4,6-tetradeoxy-D-arabino-hexopyranosyl]-D-chiro-inositol
Empirical Formula (Hill Notation):
C14H25N3O9 · HCl
CAS Number:
Molecular Weight:
415.82
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing