
Isazophos

Empirical Formula (Hill Notation):
C9H17ClN3O3PS
CAS Number:
Molecular Weight:
313.74
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing