
Hyperforin (stable Dicyclohexylammonium salt)

Synonym(s):
Hyperforin (dicyclohexylammonium) salt, Hyp-DCHA, Hyperforin-DCHA
Empirical Formula (Hill Notation):
C35H52O4·C12H23N
CAS Number:
Molecular Weight:
718.10
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing