
Hydroxylammonium chloride

Synonym(s):
Hydroxylammonium chloride
Empirical Formula (Hill Notation):
H3NO · xHCl
CAS Number:
Molecular Weight:
33.03 (free base basis)
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing