Skip to Content
Merck
Holmium foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99%

Holmium

Holmium foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99%
Synonym(s):
Holmium
Empirical Formula (Hill Notation):
Ho
CAS Number:
Molecular Weight:
164.93

Products

Brand
Formula Weight
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Element
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2