
Hispidulin

Synonym(s):
4′,5,7-Trihydroxy-6-methoxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, 6-Methoxyapigenin, 6-Methylscutellarein, Dinatin, Salvitin, Scutellarein 6-methyl ether
Empirical Formula (Hill Notation):
C16H12 O6
CAS Number:
Molecular Weight:
300.26
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing