
Granisetron impurity A

Synonym(s):
2-Methyl-N-[(1R,3r,5S)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-2H-indazole-3-carboxamide
Empirical Formula (Hill Notation):
C18H24N4O
CAS Number:
Molecular Weight:
312.41
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing