
Flonicamid metabolite TFNA

Synonym(s):
4-(Trifluoromethyl)pyridine-3-carboxylic acid, 4-(Trifluoromethyl)nicotinic acid
Empirical Formula (Hill Notation):
C7H4F3NO2
CAS Number:
Molecular Weight:
191.11
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing