
Erbium

Synonym(s):
Erbium
Empirical Formula (Hill Notation):
Er
CAS Number:
Molecular Weight:
167.26
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (mm)
Element
Showing 1-15 of 15
Compare
Product Number
Description
Pricing