Skip to Content
BI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Duran
®
Gls 80 Laboratory Glass Bottle Protect
Products
Product Category
bottles (3)
Capacity (mL)
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
DWK218656958
Duran
®
Gls 80 Laboratory Glass Bottle Protect,
wide-neck, plastic-coated glass bottle (PU), with screw cap to drain contents (Without pouring ring), capacity 3500 mL
Expand
View Pricing
DWK218653652
Duran
®
Gls 80 Laboratory Glass Bottle Protect,
wide-neck, plastic-coated glass bottle (PU), with screw cap to drain contents (Without pouring ring), capacity 250 mL
Expand
View Pricing
DWK218665432
Duran
®
Gls 80 Laboratory Glass Bottle Protect,
plastic-coated glass bottle (PU), amber bottle, with screw cap to drain contents: no (Without pouring ring), capacity 1000 mL
Expand
View Pricing