
Dioscin

Synonym(s):
Collettiside III, Diosgenin bis-α-L-rhamnopyranosyl-(1→2,1→4)-beta-D-glucopyranoside, Polyphyllin III
Empirical Formula (Hill Notation):
C45H72O16
CAS Number:
Molecular Weight:
869.04
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing