
Desmethoxyyangonin

Synonym(s):
(E)-4-Methoxy-6-(2-phenylethenyl)-2H-pyran-2-one, 5,6-Dehydrokavain, Demethoxyyangonin
Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing