
(+)-Catechin

Synonym(s):
(+)-Cyanidanol, D-Catechin, (+)-Catechin
Empirical Formula (Hill Notation):
C15H14O6
CAS Number:
Molecular Weight:
290.27
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Agency
Format
Quality Segment
Application
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4