![Cyproconazol PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/210/230/f76eabe6-00e2-4a59-a8d2-ed995a0c4214/640/f76eabe6-00e2-4a59-a8d2-ed995a0c4214.png)
Cyproconazol
![Cyproconazol PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/210/230/f76eabe6-00e2-4a59-a8d2-ed995a0c4214/640/f76eabe6-00e2-4a59-a8d2-ed995a0c4214.png)
Empirical Formula (Hill Notation):
C15H18ClN3O
CAS Number:
Molecular Weight:
291.78
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing