
Chlorodiisopropyloctylsilane

Synonym(s):
Diisopropyloctylchlorosilane, Octyldiisopropylchlorosilane
Linear Formula:
CH3(CH2)7Si[CH(CH3)2]2Cl
CAS Number:
Molecular Weight:
262.93
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing