Skip to Content
Merck
Cerium(IV) oxide-gadolinium doped nanopowder, contains 10 mol % gadolinium as dopant

Cerium(IV) oxide-gadolinium doped

Cerium(IV) oxide-gadolinium doped nanopowder, contains 10 mol % gadolinium as dopant
Synonym(s):
Gadolinium doped ceria
Empirical Formula (Hill Notation):
CeO2
Molecular Weight:
172.11

Products

Product Category
Sustainability
Feature
Formula Weight
Physical Form
Chemical Composition
Particle Size (µm)
Reagent Type
Core