Skip to Content
Merck
Cadusafos certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Cadusafos

Cadusafos certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
O-Ethyl S,S-bis(1-methylpropyl) phosphorodithioate
Empirical Formula (Hill Notation):
C10H23O2PS2
CAS Number:
Molecular Weight:
270.39

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique